missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EphA2/EphA3/EphA4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15567521
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15567521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567521 Supplier Invitrogen™ Supplier No. PA536108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 112 kDa. Purity is >95% by SDS-PAGE.

The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA2/EphA3/EphA4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene EPHA2
Gene Accession No. O08680, P29317, P29319, P29320, P54764, Q03137, Q03145
Gene Alias 2900005C20Rik; AI385584; ARCC2; AW492086; AW545284; Cek4; Cek8; CTPA; CTPP1; CTRCT6; ECK; EK4; EK8; End3; EPH receptor A2; Eph receptor A3; EPH receptor A4; EPHA2; EPHA3; EPHA4; EPH-like kinase 4; EPH-like kinase 8; eph-like tyrosine kinase 1; Ephrin A2; Ephrin A3; Ephrin A4; ephrin receptor EphA2; ephrin type-A receptor 2; Ephrin type-A receptor 3; ephrin type-A receptor 4; EphrinA2; EphrinA3; EphrinA4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; ETK; ETK1; HEK; HEK4; Hek8; Human embryo kinase; human embryo kinase 1; Mek4; Myk2; rb; receptor protein-tyrosine kinase HEK8; rEK4; RGD1560587; SEK; Sek1; Sek2; Sek-2; soluble EPHA2 variant 1; testicular tissue protein Li 64; Tyro1; TYRO1 protein tyrosine kinase; Tyro4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO1; tyrosine-protein kinase TYRO4
Gene Symbols EPHA2, EPHA3, EPHA4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 550-600 of human EphA2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13836, 13837, 13838, 1969, 2042, 2043, 29210, 316539, 366492
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.