missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Antibody (M060), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23320
This item is not returnable.
View return policy
Description
EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| EphA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| M060 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Clone (M060) was generated from a recombinant protein that included the extracellular region of human EphA2 protein. [UniProt# P29317] | |
| 0.1 mL | |
| Cancer, Protein Kinase | |
| 1969 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction