missing translation for 'onlineSavingsMsg'
Learn More

EphA2 Antibody (301), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18328720 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328720 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328720 Supplier Novus Biologicals Supplier No. NBP290650JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EphA2 Monoclonal specifically detects EphA2 in Mouse samples. It is validated for Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA2
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 301
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, ELISA
Formulation 50 mM Sodium Borate
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 (NP_034269.2; Met1-Asn535).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Test Specificity No cross-reactivity in ELISA with: Mouse EPHA4 Mouse EphA6 Mouse EPHA7 Human MEPHB1 Mouse EPHB3 Mouse EPHB4
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.