missing translation for 'onlineSavingsMsg'
Learn More

EphA2 Antibody (101), PE, Novus Biologicals™

Product Code. 18321011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321011 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321011 Supplier Novus Biologicals Supplier No. NBP290649PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EphA2 Monoclonal specifically detects EphA2 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA2
Applications Flow Cytometry
Classification Monoclonal
Clone 101
Conjugate PE
Formulation PBS
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 (NP_034269.2; Met1-Asn535).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.