missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EphA10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12838922
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838922 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838922 Supplier Invitrogen™ Supplier No. PA520775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20775 can be used with blocking peptide PEP-0889.

EPHA10 is a receptor for members of the ephrin-A family. It binds to EFNA3, EFNA4 and EFNA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene EPHA10
Gene Accession No. Q5JZY3, Q8BYG9
Gene Alias A330090H18; EPH receptor A10; EPHA10; EphA10s protein; ephrin type-A receptor 10; RGD1562679
Gene Symbols EPHA10
Host Species Rabbit
Immunogen A 14 amino acid peptide of near the amino terminus of human EphA10.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230735, 284656, 298528
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.