missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76318-0.025mg
This item is not returnable.
View return policy
Description
EphA10 Polyclonal specifically detects EphA10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| EphA10 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.10.1, EPH receptor A10, EphA10s protein, ephrin type-A receptor 10, FLJ16103, FLJ33655, MGC43817 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_001092909 | |
| EPHA10 | |
| Antibody was raised against a 14 amino acid synthetic peptide of near the amino terminus of human EphA10. The immunogen is located within amino acids 120 - 170 of EphA10. Amino Acid Squence: RPRLGGSRPRKIDT | |
| 0.025 mg | |
| Protein Kinase | |
| 284656 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction