missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA1 Rabbit anti-Mouse, Clone: 43, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90732-100UL
This item is not returnable.
View return policy
Description
EphA1 Monoclonal specifically detects EphA1 in Mouse samples. It is validated for ELISA.
Specifications
| EphA1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 43 | |
| ELISA 1:5000-1:10000 | |
| EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH | |
| Recombinant Mouse EphA1 protein | |
| 100 μg | |
| Protein Kinase | |
| 2041 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction