missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epha1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547435
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 35% cross-reactivity with recombinant human EphA1 and less than 5% cross-reactivity with recombinant mouse (rm) EphA4, rmEphA6, and rmEphA7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
EphA1 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA1 is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
Specifications
| Epha1 | |
| Polyclonal | |
| Unconjugated | |
| EPHA1 | |
| 5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; EPH receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; tyrosine-protein kinase receptor EPH; Tyrosine-protein kinase receptor ESK | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 13835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q60750 | |
| EPHA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA1 Glu27-Glu548. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction