missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA1 Mouse anti-Human, Clone: 1B7C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60155-2-IG-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis.Specifications
| EPHA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21709 | |
| EPHA1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1B7C3 | |
| Unconjugated | |
| EPHA1 | |
| EPH, EPH receptor A1, EPHA1, Ephrin type A receptor 1, EPHT, EPHT1 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 2041 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction