missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphA1 Monoclonal Antibody (404517)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561248
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
EphA1 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA1 is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
Specifications
| EphA1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q60750 | |
| EPHA1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA1, Glu27-Glu548. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 404517 | |
| Unconjugated | |
| EPHA1 | |
| 5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; EPH receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; tyrosine-protein kinase receptor EPH; Tyrosine-protein kinase receptor ESK | |
| Rat | |
| Protein A/G | |
| RUO | |
| 13835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction