missing translation for 'onlineSavingsMsg'
Learn More

EphA1 Antibody (404517) [Allophycocyanin], Novus Biologicals™

Product Code. 30735161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735161 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735161 Supplier Novus Biologicals Supplier No. FAB30341A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

EphA1 Monoclonal antibody specifically detects EphA1 in Mouse samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA1
Applications Immunocytochemistry
Classification Monoclonal
Clone 404517
Conjugate APC
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA1, Glu27-Glu548, Accession # AAH71215
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2041
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.