missing translation for 'onlineSavingsMsg'
Learn More

EphA1 Antibody (043), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18331701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331701 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331701 Supplier Novus Biologicals Supplier No. NBP290732AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EphA1 Monoclonal specifically detects EphA1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA1
Applications ELISA
Classification Monoclonal
Clone 43
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA1 (Q60750; Met1-Glu548).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2041
Test Specificity No cross-reactivity in ELISA with: Mouse EphA6 Mouse EPHA7 Mouse EPHB3 Mouse EPHB4
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.