missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPG5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538062
This item is not returnable.
View return policy
Description
Do not aliquot.
Specifications
| EPG5 | |
| Polyclonal | |
| Unconjugated | |
| EPG5 | |
| Ectopic P granules autophage protein 5 homolog antibody, HEEW1 antibody, hEPG5 antibody | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100502841, 364902, 57724 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HCE0, Q80TA9 | |
| EPG5 | |
| A synthetic peptide derived from human EPG5 (241-331aa) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction