missing translation for 'onlineSavingsMsg'
Learn More

EPDR1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18324303 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324303 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324303 Supplier Novus Biologicals Supplier No. NBP300177G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPDR1 Polyclonal specifically detects EPDR1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPDR1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EPDR, ependymin related protein 1 (zebrafish), MERP1mammalian ependymin related protein 1, MERP-1Upregulated in colorectal cancer gene 1 protein, UCC1mammalian ependymin-related protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EPDR1 (Q9UM22-1; Met1-Trp224).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54749
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.