missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11391-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregcy.Specifications
| EPCR | |
| Polyclonal | |
| Unconjugated | |
| PROCR | |
| Activated protein C receptor, APC receptor, CCCA, CCD41, CD201, Endothelial protein C receptor, EPCR, PROCR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10544 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UNN8 | |
| PROCR | |
| EPCR Fusion Protein Ag1937 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction