missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPCR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100503
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EPCR.
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
Specifications
| EPCR | |
| Polyclonal | |
| Unconjugated | |
| Procr | |
| Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10544, 19124, 362248 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4V8I1, Q64695, Q9UNN8 | |
| Procr | |
| A synthesized peptide derived from human PROCR(Accession Q9UNN8), corresponding to amino acid residues T174-G224. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction