missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPCR Monoclonal Antibody (RCR-252), FITC
Rat Monoclonal Antibody
Brand: Invitrogen™ MA549090
This item is not returnable.
View return policy
Description
EPCR Monoclonal Antibody for Western Blot, Flow, IP, Functional Assay
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
Specifications
| EPCR | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.02% sodium azide | |
| Q9UNN8 | |
| Procr | |
| Human EPCR-positive RE-1 cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
| RCR-252 | |
| FITC | |
| Procr | |
| Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
| Rat | |
| Protein G | |
| RUO | |
| 10544 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction