missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR (CD201) Rat, anti-Mouse, FITC, Clone: RMEPCR1560 (1560), STEMCELL Technologies
Rat Monoclonal Antibody
POA - £233.55
Specifications
| Antigen | EPCR (CD201) |
|---|---|
| Clone | RMEPCR1560 (1560) |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178701
|
Stemcell Technologies
60038FI.1 |
25 Tests |
£233.55
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168701
|
Stemcell Technologies
60038FI |
100 Tests | |||||||
Description
- The RMEPCR1560 (1560) antibody reacts with the endothelial protein C receptor (EPCR or CD201), an ∼25 kDa type I transmembrane glycoprotein expressed by endothelial cells, subsets of hematopoietic stem cells (HSCs) and dendritic cells, and several malignant cell lines
- It is also found in a soluble form in plasma
- EPCR exhibits homology with the MHC class 1/CD1 protein family
- EPCR binds protein C and activated protein C, thus augmenting protein C activation by the thrombin-thrombomodulin complex and regulating blood coagulation and inflammation
- EPCR protein expression has been detected on ∼1.5% of mouse bone marrow cells
- Purified EPCR+ cells are highly enriched for HSC activity, as evidenced by high in vivo repopulation activity
- Moreover, EPCR expression is associated with the stem cell activity of bone marrow cell populations isolated using conventional markers, indicating the usefulness of EPCR as a single marker for the identification of mouse HSCs
Specifications
| EPCR (CD201) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Functional Assay | |
| FITC | |
| Mouse | |
| Primary |
| RMEPCR1560 (1560) | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title