missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR Antibody (OTI12H5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71607CL1
This item is not returnable.
View return policy
Description
EPCR Monoclonal antibody specifically detects EPCR in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| EPCR | |
| Monoclonal | |
| CoraFluor 1 | |
| APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial | |
| Human recombinant protein fragment corresponding to amino acids 1-210 of human PROCR (NP_006395) produced in HEK293T cell. | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 10544 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI12H5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction