missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EPCR Monoclonal antibody specifically detects EPCR in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Conjugate | mFluor Violet 450 SE |
| Dilution | Immunohistochemistry-Paraffin |
| Gene Alias | APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human EPCR (Uniprot#: Q9UNN8; Met1-Thr209). |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?