missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Rabbit anti-Mouse, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £458.00
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Clone | 2 |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000, Sandwich ELISA Detection 1:5000-1:50000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267577
|
Novus Biologicals
NBP2-89589-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18255848
|
Novus Biologicals
NBP2-89589-100UL |
100 μg |
£458.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.Specifications
| EpCAM/TROP1 | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000, Sandwich ELISA Detection 1:5000-1:50000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| Recombinant Mouse EpCAM/TROP1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cancer Stem Cells | |
| 0.2 um filtered solution in PBS | |
| 4072 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title