missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody (rVU-1D9) - (ECD), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £409.00
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Clone | rVU-1D9 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109829
|
Novus Biologicals
NBP2-53265-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237830
|
Novus Biologicals
NBP2-53265-100UG |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human, Ferret (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EpCAM/TROP1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cancer Stem Cells | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 4072 | |
| Small cell lung carcinoma cells | |
| Primary | |
| Store at 4C. |
| rVU-1D9 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Ferret (Negative), Rat (Negative) | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| TACSTD1 | |
| IgG1 κ | |
| Protein A or G purified | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title