missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody (EGP40/1798), FITC - (ECD), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54456F
This item is not returnable.
View return policy
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human, Canine, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| EpCAM/TROP1 | |
| Monoclonal | |
| FITC | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| Recombinant fragment from the extracellular domain of human EpCAM/TROP1 protein (around aa100-224) (exact sequence is proprietary) (Uniprot: P16422 ) | |
| 100 μg | |
| Cancer, Cancer Stem Cells, Tumor Biomarkers | |
| 4072 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| EGP40/1798 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction