missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody (EGP40/1555R), DyLight 350 - (ECD), Novus Biologicals™
Shop All Bio Techne ProductsDescription
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | EpCAM/TROP1 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | EGP40/1555R |
| Conjugate | DyLight 350 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A |
| Host Species | Rabbit |
| Immunogen | Recombinant full-length human EpCAM/TROP1 protein (Uniprot: P16422 ) |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?