missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCAM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21050-1-AP-150UL
This item is not returnable.
View return policy
Description
Epithelial cell adhesion molecule (EpCAM) is a transmembrane glycoprotein mediating Ca2+-independent homotypic cell-cell adhesion in epithelia. This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy.Specifications
| EPCAM | |
| Polyclonal | |
| Unconjugated | |
| EPCAM | |
| CD326, CO 17A, CO17 1A, EGP, EGP 2, EGP314, EGP34, EGP40, Ep CAM, EPCAM, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733 2, hEGP 2, hEGP314, KS 1/4 antigen, KS1/4, KSA, M1S2, M4S1, MIC18, MK 1, TACST 1, TACSTD1, TROP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17075, 4072 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16422, Q99JW5 | |
| EPCAM | |
| EPCAM Fusion Protein Ag15393 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction