missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM Monoclonal Antibody (VU-1D9), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512150
This item is not returnable.
View return policy
Description
MA5-12150 targets Epithelial Specific Antigen in IHC (P) applications and shows reactivity with Human samples. The MA5-12150 immunogen is small cell lung carcinoma cells.
EpCAM Monoclonal antibody specifically detects EpCAM in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
| EpCAM | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P16422 | |
| EPCAM | |
| Small cell lung carcinoma cells | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| VU-1D9 | |
| Biotin | |
| EPCAM | |
| adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 4072 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction