missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epcam / CD326 Rabbit, Clone: EGP40/1556R, enQuireBio™
Rabbit Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | Epcam / CD326 |
|---|---|
| Clone | EGP40/1556R |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| Epcam / CD326 | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 4072 | |
| Recombinant human EPCAM protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 40-43kDa |
| EGP40/1556R | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_002345 | |
| EPCAM | |
| IgG κ | |
| Protein A/G Purified | |
| Cell surface | |
| HT29 cells or breast tumor |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title