missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71865
This item is not returnable.
View return policy
Description
EPC1 Polyclonal specifically detects EPC1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| EPC1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp781P2312, enhancer of polycomb 1, enhancer of polycomb homolog 1, enhancer of polycomb homolog 1 (Drosophila), Epl1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80314 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
| NP_079485.1 | |
| EPC1 | |
| The immunogen for this product maps to a region between residue 325 and 375 of human Enhancer of Polycomb 1 using the numbering given in entry NP_079485.1 (GeneID 80314). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction