missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71865AF647
This item is not returnable.
View return policy
Description
EPC1 Polyclonal antibody specifically detects EPC1 in Human samples. It is validated for Western Blot
Specifications
| EPC1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp781P2312, enhancer of polycomb 1, enhancer of polycomb homolog 1, enhancer of polycomb homolog 1 (Drosophila), Epl1 | |
| The immunogen for this product maps to a region between residue 325 and 375 of human Enhancer of Polycomb 1 using the numbering given in entry NP_079485.1 (GeneID 80314). | |
| 0.1 mL | |
| modENCODE Antibodies | |
| 80314 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction