missing translation for 'onlineSavingsMsg'
Learn More

EPC1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18415483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415483 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415483 Supplier Novus Biologicals Supplier No. NBP171865AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPC1 Polyclonal antibody specifically detects EPC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPC1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp781P2312, enhancer of polycomb 1, enhancer of polycomb homolog 1, enhancer of polycomb homolog 1 (Drosophila), Epl1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 325 and 375 of human Enhancer of Polycomb 1 using the numbering given in entry NP_079485.1 (GeneID 80314).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 80314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.