missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB4IL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33426
This item is not returnable.
View return policy
Description
EPB4IL2 Polyclonal specifically detects EPB4IL2 in Human samples. It is validated for Western Blot.
Specifications
| EPB4IL2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| EPB41L2 | |
| Recombinant protein encompassing a sequence within the center region of human EPB41L2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2037 | |
| Human | |
| IgG |
| Western Blot | |
| 0.62mg/mL | |
| Western Blot 1:500-1:3000 | |
| band 4.1-like protein 2,4.1G, DKFZp781D1972, DKFZp781H1755,4.1-G, erythrocyte membrane protein band 4.1 like-protein 2, erythrocyte membrane protein band 4.1-like 2, Generally expressed protein 4.1 | |
| Rabbit | |
| 113 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction