missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPB42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559127
This item is not returnable.
View return policy
Description
Immunogen sequence: FAQEDIAICR PHLAIKMPEK AEQYQPLTAS VSLQNSLDAP MEDCVISILG RGLIHRERSY RFRSVWPENT MCAKFQFTPT HVGLQRLTVE VDCNMFQNLT NYKSVT Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 68%.
Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| EPB42 | |
| Polyclonal | |
| Unconjugated | |
| EPB42 | |
| BEP42; E42P; Epb4.2; Epb42; Erythrocyte membrane protein band 4.2; erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; membrane skeletal protein; P4.2; PA; SPH5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2038 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16452 | |
| EPB42 | |
| Recombinant protein corresponding to Human EPB42. Recombinant protein control fragment (Product #RP-98173). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction