missing translation for 'onlineSavingsMsg'
Learn More

EPB42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen EPB42
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18251275
View Documents
Novus Biologicals
NBP1-56647
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EPB42 Polyclonal specifically detects EPB42 in Human samples. It is validated for Western Blot.
Specifications

Specifications

EPB42
Polyclonal
Rabbit
P16452-2
2038
Synthetic peptides corresponding to EPB42 (erythrocyte membrane protein band 4.2) The peptide sequence was selected from the middle region of EPB42. Peptide sequence ISTKGVGSDRCEDITQNYKYPEGSLQEKEVLERVEKEKMEREKDNGIRPP.
Primary
Western Blot
Unconjugated
RUO
E42P, erythrocyte membrane protein band 4.2, Erythrocyte protein 4.2, erythrocyte surface protein band 4.2, MGC116735, MGC116737, P4.2, PA, SPH5
EPB42
IgG
80 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.