missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41L5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £444.00
Specifications
| Antigen | EPB41L5 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696515
|
Novus Biologicals
NBP2-38354-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18091714
|
Novus Biologicals
NBP2-38354 |
0.1 mL |
£444.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EPB41L5 Polyclonal specifically detects EPB41L5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| EPB41L5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9HCM4 | |
| 57669 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DLLEATIGDVIGASDTMETSQALNDVNVATRLPGLGEPEVEYETLKDTSEKLKQLEMENSPLLSPRSNIDVNINSQEEVVKLTEKC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| band 4.1-like protein 5, erythrocyte membrane protein band 4.1 like 5, FLJ12957, KIAA1548BE37, YMO1 | |
| EPB41L5 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title