missing translation for 'onlineSavingsMsg'
Learn More

EPB41L4A Antibody, Novus Biologicals™

Product Code. 18354891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354891 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354891 Supplier Novus Biologicals Supplier No. NBP287367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPB41L4A Polyclonal specifically detects EPB41L4A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L4A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias DKFZp566L203, EPB41L4erythrocyte protein band 4.1-like 4, erythrocyte membrane protein band 4.1 like 4A, FLJ38738, NBL4band 4.1-like protein 4A, Protein NBL4
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human EPB41L4A. Peptide sequence: QPVRRRKAHNSGEDSDLKQRRRSRSRCNTSSGSESENSNREHRKKRNRI The peptide sequence for this immunogen was taken from within the described region.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64097
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.