missing translation for 'onlineSavingsMsg'
Learn More

EPB41L3 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18298910 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18298910 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18298910 Supplier Novus Biologicals Supplier No. NB11061027V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPB41L3 Polyclonal specifically detects EPB41L3 in Human, Mouse, Rat, Primate, Xenopus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPB41L3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9Y2J2
Gene Alias 4.1BFLJ37633, DAL1DAL-1,, Differentially expressed in adenocarcinoma of the lung protein 1, erythrocyte membrane protein band 4.1-like 3, KIAA0987band 4.1-like protein 3
Gene Symbols EPB41L3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500). [Swiss-Prot# Q9Y2J2]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 23136
Target Species Human, Mouse, Rat, Primate, Xenopus species
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.