Learn More
EPB41L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563352
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KKAQEEAPQQ PEAAAAVTTP VTPAGHGHPE ANSNEKHPSQ QDTRPAEQSL DMEEKDYSEA DGLSERTTPS KAQKSPQKIA K Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane.Specifications
EPB41L1 | |
Polyclonal | |
Unconjugated | |
EPB41L1 | |
4.1N; band 4.1-like protein 1; DKFZp686H17242; Epb4; Epb4.1l1; Epb41l1; erythrocyte membrane protein band 4.1 like 1; erythrocyte membrane protein band 4.1-like 1; erythrocyte protein band 4.1 like 1; erythrocyte protein band 4.1-like 1; KIAA0338; MGC11072; mKIAA0338; MRD11; NBL1; neuronal protein 4.1; neuron-type nonerythroid protein 4.1; RP11-234K24.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4G0 | |
EPB41L1 | |
Recombinant protein corresponding to Human EPB41L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only