Learn More
EPB41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542239
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SESRGLSRLF SSFLKRPKSQ VSEEEGKEVE SDKEKGEGGQ KEIEFGTSLD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
EPB41 Polyclonal antibody specifically detects EPB41 in Human samples. It is validated for Western Blot.Specifications
EPB41 | |
Polyclonal | |
Unconjugated | |
EPB41 | |
4.1R; AI415518; Band 4.1; D4Ertd442e; E41P; EL1; elliptocytosis 1, RH-linked; Elp1; Elp-1; EPB4.1; EPB41; erythrocyte membrane protein band 4.1; erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked); erythrocyte protein band 4.1; erythrocyte skeletal protein; erythrocyte surface protein band 4.1; HE; Kiaa4056; mKIAA4056; P4.1; protein 4.1; Protein 4.1R; RGD1564762 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2035 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P11171 | |
EPB41 | |
Synthetic peptide directed towards the N-terminal of human EPB41 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |