missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPB41 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37997-100ul
This item is not returnable.
View return policy
Description
EPB41 Polyclonal antibody specifically detects EPB41 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EPB41 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| 4.1RBand 4.1, E41P, EL1, EPB4.1, erythrocyte membrane protein band 4.1 (elliptocytosis 1, RH-linked), erythrocyte surface protein band 4.1, HE, P4.1, protein 4.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-500 of human EPB41 (NP_001159478.1).,, Sequence:, MHCKVSLLDDTVYECVVEKHAKGQDLLKRVCEHLNLLEEDYFGLAIWDNATSKTWLDSAKEIKKQVRGVPWNFTFNVKFYPPDPAQLTEDITRYYLCLQLRQDIVAGRLPCSFATLALLGSYTIQSELGDYDPELHGVDYVSDFKLAPNQTKELEEKVMELHKSYRSMTPAQADLEFLENAKKLSMYGVDLHKAKDLEGVDIILGVCSSGLLVYKDKLRINRFPWPKVLKISYKRSSFFIKIRPGEQEQYESTIGFKLPSYRAAKKLWKVCVEHHTFFRLTSTDTIPKSKF | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2035 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction