missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
Specifications
Specifications
| Antigen | EPAS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2F6B12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | EPAS1 |
| Gene Accession No. | Q99814 |
| Gene Alias | bHLHe73, ECYT4, EPAS 1, EPAS1, HIF 1 alpha like factor, HIF 2 alpha, HIF2 alpha, HIF2A, HLF, Member of PAS protein 2, MOP2, PASD2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?