missing translation for 'onlineSavingsMsg'
Learn More

Epacadostat, TRC

Product Code. 30371812
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371812 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371812 Supplier Toronto Research Chemicals Supplier No. TRCE56550350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epacadostat
CAS 1204669-58-8
Molecular Formula C11 H13 Br F N7 O4 S
Quantity 50 mg
Synonym 1,2,5-Oxadiazole-3-carboximidamide, 4-[[2-[(aminosulfonyl)amino]ethyl]amino]-N-(3-bromo-4-fluorophenyl)-N'-hydroxy-, [C(Z)]-, [C(Z)]-4-[[2-[(Aminosulfonyl)amino]ethyl]amino]-N-(3-bromo-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide, Epacadostat, ICBN 24360, INCB 024360
InChI Formula InChI=1S/C11H13BrFN7O4S/c12-7-5-6(1-2-8(7)13)17-11(18-21)9-10(20-24-19-9)15-3-4-16-25(14,22)23/h1-2,5,16,21H,3-4H2,(H,15,20)(H,17,18)(H2,14,22,23)
SMILES NS(=O)(=O)NCCNc1nonc1\C(=N\O)\Nc2ccc(F)c(Br)c2
IUPAC Name N-(3-bromo-4-fluorophenyl)-N'-hydroxy-4-[2-(sulfamoylamino)ethylamino]-1,2,5-oxadiazole-3-carboximidamide
Molecular Weight (g/mol) 438.23
Formula Weight 436.9917
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.