missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPAC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19103-1-AP-150UL
This item is not returnable.
View return policy
Description
Guanine nucleotide exchange factor (GEF) for RAP1A, RAP1B and RAP2A small GTPases that is activated by binding cAMP. Seems not to activate RAB3A. Involved in cAMP-dependent, PKA-independent exocytosis through interaction with RIMS2.Specifications
| EPAC2 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF4 | |
| CAMP GEFII, CGEF2, EPAC 2, EPAC2, Nbla00496, RAPGEF4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11069, 252857, 56508 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WZA2, Q9EQZ6, Q9Z1C7 | |
| Rapgef4 | |
| EPAC2 Fusion Protein Ag5186 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction