missing translation for 'onlineSavingsMsg'
Learn More

Epac2 Antibody (1C11), Novus Biologicals™

Product Code. 18379969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379969 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379969 Supplier Novus Biologicals Supplier No. H00011069M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Epac2 Monoclonal antibody specifically detects Epac2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epac2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH24004
Gene Alias CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4
Host Species Mouse
Immunogen RAPGEF4 (AAH24004, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVAAHAAHSSSSAEWIACLDKRPLERSSEDVDIIFTRLKEVKAFEKFHPNLLHQICLCGYYENLEKGITLFRQGDIGTNWYAVLAGSLDVKVSETSSHQDAVTICTLGIG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11069
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.