missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 Nitrosamines Mixture 339 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 4-Nitrosomorpholine∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| CAS | 59-89-2, 55-18-5, 924-16-3, 10595-95-6, 100-75-4, 930-55-2 |
| Analyte or Component Names | 4-Nitrosomorpholine∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 4-Nitrosomorpholine 2000 μg/mL∣N-Nitroso-diethylamine 2000 μg/mL∣N-Nitroso-di-n-butylamine 2000 μg/mL∣N-Nitrosomethylethylamine 2000 μg/mL∣N-Nitrosopiperidine 2000 μg/mL∣N-Nitrosopyrrolidine 2000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?