missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 Calibration by Class Mixture 4 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1,3,5-Trinitrobenzene∣1,4-Naphthoquinone∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣4-Nitroquinoline-1-oxide∣Acetophenone∣Azobenzene∣Benzyl alcohol∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Ethyl methanesulfonate∣Isophorone∣Isosafrole∣Methylmethanesulfonate∣Nitrobenzene∣Phenacetin∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Safrole |
| CAS | 99-35-4, 130-15-4, 121-14-2, 606-20-2, 56-57-5, 98-86-2 |
| Analyte or Component Names | 1,3,5-Trinitrobenzene∣1,4-Naphthoquinone∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣4-Nitroquinoline-1-oxide∣Acetophenone∣Azobenzene∣Benzyl alcohol∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Ethyl methanesulfonate∣Isophorone∣Isosafrole∣Methylmethanesulfonate∣Nitrobenzene∣Phenacetin∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Safrole |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,3,5-Trinitrobenzene 2000 μg/mL∣1,4-Naphthoquinone 2000 μg/mL∣2,4-Dinitrotoluene 2000 μg/mL∣2,6-Dinitrotoluene 2000 μg/mL∣4-Nitroquinoline-1-oxide 2000 μg/mL∣Acetophenone 2000 μg/mL∣Azobenzene 2000 μg/mL∣Benzyl alcohol 2000 μg/mL∣Dibenzofuran 2000 μg/mL∣Dibutyl phthalate 2000 μg/mL∣Diethyl phthalate 2000 μg/mL∣Di-n-octyl phthalate 2000 μg/mL∣Ethyl methanesulfonate 2000 μg/mL∣Isophorone 2000 μg/mL∣Isosafrole 2000 μg/mL∣Methylmethanesulfonate 2000 μg/mL∣Nitrobenzene 2000 μg/mL∣Phenacetin 2000 μg/mL∣Phthalic acid, benzylbutyl ester 2000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 2000 μg/mL∣Phthalic acid, bis-methyl ester 2000 μg/mL∣Safrole 2000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction