missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Calibration by Class Mixture 2 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30319537
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30319537 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30319537

Brand: Dr. Ehrenstorfer DREGA09000438DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitroaniline∣3,3'-Dichlorobenzidine∣3-Nitroaniline∣4,4'-Benzidine∣4-Chloroaniline∣4-Nitroaniline∣Aniline∣Diphenylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣Pyridine
CAS 88-74-4, 91-94-1, 99-09-2, 92-87-5, 106-47-8, 100-01-6
Analyte or Component Names 2-Nitroaniline∣3,3'-Dichlorobenzidine∣3-Nitroaniline∣4,4'-Benzidine∣4-Chloroaniline∣4-Nitroaniline∣Aniline∣Diphenylamine∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣Pyridine
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2-Nitroaniline 2000 μg/mL∣3,3'-Dichlorobenzidine 2000 μg/mL∣3-Nitroaniline 2000 μg/mL∣4,4'-Benzidine 2000 μg/mL∣4-Chloroaniline 2000 μg/mL∣4-Nitroaniline 2000 μg/mL∣Aniline 2000 μg/mL∣Diphenylamine 2000 μg/mL∣N-Nitrosodimethylamine 2000 μg/mL∣N-Nitroso-di-n-propylamine 2000 μg/mL∣Pyridine 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.