missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 B/N Surrogate Mixture 5000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30324735
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 30324735

Brand: Dr. Ehrenstorfer DRESY09000008DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluorobiphenyl∣Nitrobenzene D5∣p-Terphenyl D14
CAS 321-60-8, 4165-60-0, 1718-51-0
Analyte or Component Names 2-Fluorobiphenyl∣Nitrobenzene D5∣p-Terphenyl D14
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 5 x 5 mL
Concentration or Composition (by Analyte or Components) 2-Fluorobiphenyl 5000 μg/mL∣Nitrobenzene D5 5000 μg/mL∣p-Terphenyl D14 5000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.