missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Appendix IX Mixture 2 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30323042
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323042 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323042 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000445DI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetrachlorobenzene∣1,3,5-Trinitrobenzene∣1,4-Dioxane∣1,4-Naphthoquinone∣1-Chloronaphthalene∣2,6-Dichlorophenol∣3-Methylcholanthrene∣4-Nitroquinoline-1-oxide∣7,12-Dimethylbenzo(a)anthracene∣Acetophenone∣Aramite∣Benzaldehyde∣Biphenyl∣Chlordecone∣Chlorobenzilate∣Diallate∣Dibenz(A,J)Acridine∣e-Caprolactam∣Ethyl methanesulfonate∣Hexachloropropene (Perchloropropene)∣Isodrin∣Isosafrole∣Methacrylic acid-ethyl ester∣Methylmethanesulfonate∣Oxydibenzene∣Pentachlorobenzene∣Pentachloroethane∣Phenacetin∣Propyzamide∣Quintozene∣Safrole
CAS 95-94-3, 99-35-4, 123-91-1, 130-15-4, 90-13-1, 87-65-0
Analyte or Component Names 1,2,4,5-Tetrachlorobenzene∣1,3,5-Trinitrobenzene∣1,4-Dioxane∣1,4-Naphthoquinone∣1-Chloronaphthalene∣2,6-Dichlorophenol∣3-Methylcholanthrene∣4-Nitroquinoline-1-oxide∣7,12-Dimethylbenzo(a)anthracene∣Acetophenone∣Aramite∣Benzaldehyde∣Biphenyl∣Chlordecone∣Chlorobenzilate∣Diallate∣Dibenz(A,J)Acridine∣e-Caprolactam∣Ethyl methanesulfonate∣Hexachloropropene (Perchloropropene)∣Isodrin∣Isosafrole∣Methacrylic acid-ethyl ester∣Methylmethanesulfonate∣Oxydibenzene∣Pentachlorobenzene∣Pentachloroethane∣Phenacetin∣Propyzamide∣Quintozene∣Safrole
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4,5-Tetrachlorobenzene 1000 μg/mL∣1,3,5-Trinitrobenzene 1000 μg/mL∣1,4-Dioxane 1000 μg/mL∣1,4-Naphthoquinone 1000 μg/mL∣1-Chloronaphthalene 1000 μg/mL∣2,6-Dichlorophenol 1000 μg/mL∣3-Methylcholanthrene 1000 μg/mL∣4-Nitroquinoline-1-oxide 1000 μg/mL∣7,12-Dimethylbenzo(a)anthracene 1000 μg/mL∣Acetophenone 1000 μg/mL∣Aramite 1000 μg/mL∣Benzaldehyde 1000 μg/mL∣Biphenyl 1000 μg/mL∣Chlordecone 1000 μg/mL∣Chlorobenzilate 1000 μg/mL∣Diallate 1000 μg/mL∣Dibenz(A,J)Acridine 1000 μg/mL∣e-Caprolactam 1000 μg/mL∣Ethyl methanesulfonate 1000 μg/mL∣Hexachloropropene (Perchloropropene) 1000 μg/mL∣Isodrin 1000 μg/mL∣Isosafrole 1000 μg/mL∣Methacrylic acid-ethyl ester 1000 μg/mL∣Methylmethanesulfonate 1000 μg/mL∣Oxydibenzene 1000 μg/mL∣Pentachlorobenzene 1000 μg/mL∣Pentachloroethane 1000 μg/mL∣Phenacetin 1000 μg/mL∣Propyzamide 1000 μg/mL∣Quintozene 1000 μg/mL∣Safrole 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.