missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8015 Non-halogenated VOC Mixture 412 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,4-Dioxane∣2-Butanone∣4-Methyl-2-pentanone (MIBK)∣Acetonitrile∣Acrylamide∣Diethylether∣Ethanol∣Isobutanol∣Methacrylic acid-ethyl ester∣Methacrylic acid-methyl ester∣Methacrylonitrile∣Propionitrile |
| CAS | 123-91-1, 78-93-3, 108-10-1, 75-05-8, 79-06-1, 60-29-7 |
| Analyte or Component Names | 1,4-Dioxane∣2-Butanone∣4-Methyl-2-pentanone (MIBK)∣Acetonitrile∣Acrylamide∣Diethylether∣Ethanol∣Isobutanol∣Methacrylic acid-ethyl ester∣Methacrylic acid-methyl ester∣Methacrylonitrile∣Propionitrile |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Dioxane 100 μg/mL∣2-Butanone 100 μg/mL∣4-Methyl-2-pentanone (MIBK) 100 μg/mL∣Acetonitrile 100 μg/mL∣Acrylamide 100 μg/mL∣Diethylether 100 μg/mL∣Ethanol 100 μg/mL∣Isobutanol 100 μg/mL∣Methacrylic acid-ethyl ester 100 μg/mL∣Methacrylic acid-methyl ester 100 μg/mL∣Methacrylonitrile 100 μg/mL∣Propionitrile 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?