missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8015 Non-halogenated VOC Mixture 410 200 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30312454
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312454 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312454 Supplier Dr. Ehrenstorfer Supplier No. DREA50000410ME

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butanone∣4-Methyl-2-pentanone (MIBK)∣Diethylether∣Ethanol
CAS 78-93-3, 108-10-1, 60-29-7, 64-17-5
Analyte or Component Names 2-Butanone∣4-Methyl-2-pentanone (MIBK)∣Diethylether∣Ethanol
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2-Butanone 200 μg/mL∣4-Methyl-2-pentanone (MIBK) 200 μg/mL∣Diethylether 200 μg/mL∣Ethanol 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.